Would you like to log in again?
Chemotherapy Sharps Collectors, BD Medical
Supplier: BD Medical
300460 305603 305578 305614 305493 305613
BD305493CS 214.86 USD InStock
BD305493 CA-BD300460A BD305578 BD305603 BD305614 BD305613
Chemotherapy Sharps Collectors, BD Medical
Bins Sharps Containers
Units feature a wide entry port design for disposal of bulky chemotherapy waste
For accessories, see BD305451 series.
Collectors are designed to offer increased leak resistance. Units cannot be autoclaved. Plug cap provides a solid closure for leak resistant transit.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty