Would you like to log in again?
Sterile Syringe Convenience Trays, BD Medical
Supplier: BD Medical
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
305618 305617 309680 309701 309680 305617 309702 309701 305618 309703 309605 309702 309605
BD309702CS 157.54 USD InStock
BD309702 BD309701 CABD309680 CABD309701 CABD309702 CABD309605 BD309680 CA-BD305617 CA-BD305618 BD309605 BD309703 BD305618 BD305617
Sterile Syringe Convenience Trays, BD Medical
Syringes Syringes, Medical
These convenient trays provide sterile syringes with fully depressed stoppers in a single-direction orientation to assist in batch-preparation of syringes
The stopper is positioned at the bottom-most part of the tray to reduce manipulation
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty