Would you like to log in again?
BD Eclipse™ Needles and Syringes, BD
Supplier: BD Medical
BD Eclipse™
305781 305783 305781 305780 305780 305787 305782 305782 305787 305785 305786 305786
BD305780CS 222.51 USD InStock
BD305780 BD305782 BD305781 BD305783 CABD305780 BD305786 89042-540 CABD305781 CABD305782 CA89042-540 CABD305785 CABD305786
BD Eclipse™ Needles and Syringes, BD
Syringes Syringes, Medical
BD Eclipse™ Needles are designed with pivoting needle technology for single-handed activation
For use with BD Luer-Lok™ Syringes.
No hard surface is required with this needle-based, safety-engineered injection device.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty