Would you like to log in again?
BD Enteral Syringes, BD
Supplier: BD Medical
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
305857 305863 305855 305851
BD305863CS 416.97 USD InStock
BD305863 BD305851 BD305855 BD305857
BD Enteral Syringes, BD
Syringes Syringes, Medical
BD Enteral syringes offer an amber colored plunger rod and unique non-luer BD UniVia™ connection.
  • Amber-colored plunger rod eases oral delivery in low-light conditions and provides contrast between medicine and barrel
  • Clear barrel provides simple view of medication
  • Bold print makes reading scale markings easy
  • Non-luer compatible BD UniVia™ connection designed to reduce risk of tubing set misconnection
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty