Would you like to log in again?
Alcohol Swabs, BD™
Supplier: BD Medical
326895
BD326895CS 75.61 USD InStock
BD326895
Alcohol Swabs, BD™
Wipes Disposable Wipes and Towels Disinfectant Wipes
These thick, soft alcohol swabs ensure reliable site preparation for safe, hygienic insulin injections.
Individually wrapped in a four-layer foil wrapper to prevent leaking and drying out. Made with 70% isopropyl alcohol for optimum anti-bacterial action.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty