Would you like to log in again?
Corning® Human Umbilical Vein Endothelial Cells, HUVEC-2 Cells, Corning
Supplier: Corning
Corning®
354151 354151
BD354151EA 400.44 USD InStock
BD354151 CACB354151
Corning® Human Umbilical Vein Endothelial Cells, HUVEC-2 Cells, Corning
Cells
HUVEC-2 cells are qualified for use in Corning® BioCoat™angiogenesis systems: endothelial cell migration
The cells may be used in Corning BioCoat endothelial cell invasion and tube formation assays. They are used as model for studying endothelial differentiation and modulation of endothelial tube formation by antiangiogenic agents.
Ordering information: Includes Corning®Human Umbilical Vein Endothelial Cells, HUVEC-2 Cells (>5×105 Cells).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty