Would you like to log in again?
BD Vacutainer® Venous Blood Collection Tubes, BD Medical
Supplier: BD Medical
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
BD Vacutainer®
366430 367963 367884 367844 367841 366430 367001 368660 367844 366668 368861 367861 367871 367921 363083 367988 367977 367878 367986 366643 367820 367820 367815 367812 367962 366667 368056 367812 367921 367842 363083 367960 367814 367964 367878 363080 367729 367863 367856 367899 362788 362800 367841 366450 367874 368661 367960 367985 366480 367856 367814 367983 367964 367987 367989 367874 367922 367835 367961 367671 367899 367861 367862 367986 367962 362799 367587 367886 366664 367001 367815 367987 367878 367985 367880 366643 367871 367925 367961 367977 367880 367671 367983 367925 366408 367863 366480 367884 367922 366668 366664 366408 367988 363080 367587 367886
VT-367844CS 1588.25 USD InStock
VT-367844 BD367729 BD367922 CABD367587 BD367921 CABD367820 BD367964 CABD367985 BD367963 BD367842 BD367886 BD-366643 BD367841 CABD367987 BD367962 VT367925 BD367884 VT6480 CABD367899L CABD367863L CABD367871 CABD367815L CABD367988L VT-367001 97000-582 VT6430 10185-049 BD367815 BD367814 95026-688 BD367812 BD367856 BD367977 BD367899 CABD367880L BD-367863 CABD367977 CABD367814 CABD367844L CABD367921L VT367587 CA97000-582 CABD367812L CABD367856L CABD367925L CABD366643L BD367989 BD367988 CABD367961 CABD367884 CABD367962 BD367987 BD367986 CABD367886 BD367820 BD367985 CABD367964 CABD363083L BD367983 BD367862 CABD367922 BD367861 BD367671 CABD367841L CABD367986L CABD367861L CABD368861 BD-367878 95026-686 95045-254 CABD367671A CABD366480 CA-BD366668 CABD366664 BD367835 BD366667 BD367878 CABD367874 BD366668 BD367874 BD366664 VT362788 VT6450 BD367871 BD363083 VT367880 BD368056 VT367961 CABD367983L CABD367960 BD363080 VT367960 CABD366430 CABD367001 CABD367878L CABD363080
BD Vacutainer® Venous Blood Collection Tubes, BD Medical
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Sterile vacuum blood collection BD Vacutainer® tubes are for serum determination
Tubes with a BD Hemogard™ closure incorporate a plastic shield that protects lab personnel from contact with blood on the stopper or around the outer rim of the tube, and from potential splattering when tube is opened. The closure's rubber stopper is recessed inside the plastic shield to isolate blood drops left by the needle.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty