Would you like to log in again?
BD Vacutainer® Safety-Lok® Blood Collection Sets, BD Diagnostics
Supplier: BD Medical
BD Vacutainer®
368653 367285 367285 367294 367281 367298 368653 367297 368652 367296 367287 367297 367283 367294 367287 367283 367281 368652 367292 367296 367298 367292
BD-367296CS 533.36 USD InStock
BD-367296 CABD367281L CABD367283L CABD367285L BD367287 BD367298 CABD367292 BD367297 BD367285 89005-534 BD367294 89005-532 BD367283 BD367281 CABD368652 BD367292 CABD368653 CABD367298 CABD367287 CABD367297 CABD367296 CABD367294
BD Vacutainer® Safety-Lok® Blood Collection Sets, BD Diagnostics
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Blood collection sets consist of a 1.9cm (3/4") needle and a multi-sample Luer adapter, connected by a 17.8cm (7") or 30.5cm (12") length of tubing
Needles feature a Safety-Lok® protective shield to reduce needlestick risk. A translucent shield shows blood flashback clearly and locks retracted needle into place after use. Luer adapter allows specimen collection from a “closed system.” Sets are supplied sterile and are slim and easy to handle, with larger wing size for improved positioning during use. Safety-Lok® shields are designed to meet current OSHA safety standards for engineering controls. Needles are available with a pre-attached holder.
Ordering information: Additional Luer adapters available. Contact your VWR sales representative for more information.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty