Would you like to log in again?
BD Q-Syte™ Closed Luer Access Split Septum Device, BD Medical
Supplier: BD Medical
BD Q-Syte™
385100 385100
BD385100CS 639.16 USD InStock
CABD385100 BD385100
BD Q-Syte™ Closed Luer Access Split Septum Device, BD Medical
Catheters
Split septum device is designed to reduce catheter-related bloodstream infections associated with needleless IV systems
The smooth surface is easy to clean because there are no crevices or gaps to house bacteria. The simple fluid path design reduces places for microbes to grow and delivers a better flow rate. Clear housing allows for visual assessment of the fluid path. The Acacia Extension Set includes a 15cm (6”) small bore bi-extension and spin nut.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty