Would you like to log in again?
BD™ Whitacre Pencil Point Spinal Needles, BD Medical
Supplier: BD Medical
BD™
405079 405010
BD405079CS 1049 USD InStock
BD405079 BD405010
BD™ Whitacre Pencil Point Spinal Needles, BD Medical
Syringe Needles
Sterile, single-use needles feature a pencil point configuration that spreads dural fibers to minimize CSF loss and reduce the occurrence of post-procedure headaches
The needles advance straight through ligaments for on-target access and drug delivery, and make a distinct “pop” as the point penetrates the dura. High-strength stainless steel with special etching assures a smooth, sensitive feel and a low-glare hub provides fast, accurate visualization of CSF.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty