Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iodine 0.02M in 70/20/10 THF/pyridine/water (v/v) oxidizing reagent, for biosynthesis, VWR Chemicals BDH®

Supplier: VWR International
Danger

OB9911-014 OB9911-051 OB9911-001 OB9911-032 OB9911-157 OB9911-112
BDH0600-4LCS 1302.56 USD InStock
BDH0600-4L BDH0600-200L BDH0600-56L BDH0600-2.5L BDH0600-450ML BDH0600-20L
Iodine 0.02M in 70/20/10 THF/pyridine/water (v/v) oxidizing reagent, for biosynthesis, VWR Chemicals BDH®
Iodine
BDH Biosynthesis grade reagents are designed for biosynthesis research and production. A range of sizing is available to fit your needs, from benchtop synthesizers to bulk production.

  • Custom blends and bulk sizes available upon request.


Formula: I₂
MW: 253.81 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011355
CAS Number: 7553-56-2
UN: 1993
ADR: 3,II
Order Now
Specification Test Results
Description (Dark Iodine Colored Solution) Pass
Iodine 0.019-0.021 M
Pyridine 19-21%
Tetrahydrofuran 69-71%
Water 9-11%
Iodine (component purity) 99.8%
Pyridine (component purity) 99.8%
Tetrahydrofuran (component purity) 99.9%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty