Synonyms:
BTT
, 5-(Benzylthio)-1H-tetrazole
, BMT
AB3964-112
AB3964-014
AB3964-157
AB3964-032
AB3964-051
AB3964-001
BDH0601-2.5LCS
3560.84
USD
InStock
BDH0601-2.5L
BDH0601-4L
BDH0601-200L
BDH0601-450ML
BDH0601-56L
BDH0601-20L
5-Benzylmercaptotetrazole (BMT, BTT) 0.20M in anhydrous acetonitrile activator reagent, for biosynthesis, VWR Chemicals BDH®
5-(Benzylthio)-1H-tetrazole
BDH Biosynthesis Grade Reagents are designed for Biosynthesis research and production. Sizing available to fit your needs, from benchtop synthesizers to bulk production.
- Custom Blends and Bulk Sizes available upon request.