Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-(Ethylthio)-1H-tetrazole (ETT) 0.25M in anhydrous acetonitrile activator reagent, for biosynthesis, VWR Chemicals BDH®

Supplier: VWR International
Danger
Synonyms: 5-Ethyltio-1H-Tetrazole

AB3962-014 AB3962-157 AB3962-051 AB3962-032 AB3962-024 AB3962-001
BDH0602-20LCS 23124.55 USD InStock
BDH0602-20L BDH0602-450ML BDH0602-56L BDH0602-2.5L BDH0602-4L BDH0602-200L
5-(Ethylthio)-1H-tetrazole (ETT) 0.25M in anhydrous acetonitrile activator reagent, for biosynthesis, VWR Chemicals BDH®
5-(Ethylthio)-1H-tetrazole (ETT)
BDH Biosynthesis grade reagents are designed for biosynthesis research and production. sizing available to fit your needs, from benchtop synthesizers to bulk production.

  • Custom blends and bulk sizes available upon request.


Formula: C₃H₆N₄S
MW: 130.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00068733
CAS Number: 89797-68-2
UN: 1648
ADR: 3,II
Order Now
Specification Test Results
Description (Clear liquid free of suspended matter) Pass
5-Ethythio-1H-tetrazole Concentration 0.24 - 0.26 M
Color (at time of packaging) <10 APHA
Water Max 30 ppm
Acetonitrile (component purity) 99.9%
5-Ethythio-1H-tetrazole (component purity) 99.0 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty