Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Methylimidazole (NMI) 16% in anhydrous THF for biosynthesis Cap B, VWR Chemicals BDH®

Supplier: VWR International
Danger
Synonyms: N-MethylimidazoleNMI1-Methyl-1H-imidazoleN-Methyl-1H-imidazole

CB3991-157 CB3991-112 CB3991-032 CB3991-001 CB3991-014 CB3991-051
BDH0606-20LCS 9602.78 USD InStock
BDH0606-20L BDH0606-450ML BDH0606-2.5L BDH0606-56L BDH0606-4L BDH0606-200L
1-Methylimidazole (NMI) 16% in anhydrous THF for biosynthesis Cap B, VWR Chemicals BDH®
1-Methylimidazole
BDH Biosynthesis Grade Reagents are designed for Biosynthesis research and production. Sizing available to fit your needs, from benchtop synthesizers to bulk production.

  • Custom Blends and Bulk Sizes available upon request.


Formula: C₄H₆N₂
MW: 82.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005292
CAS Number: 616-47-7
UN: 2924
ADR: 3 (8),II
Order Now
Specification Test Results
Description (Clear liquid free of suspended matter) Pass
1-Methyl imidazole 14.0 - 16.0%
Tetrahydrofuran 84.0 - 86.0%
Color (at time of packaging) <10 APHA
Water Max 150 ppm
1-Methyl imidazole (component purity) Min 99.0%
Tetrahydrofuran (component purity) Min 99.9%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty