Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cap B 50:20:30 Anhydrous ACN/Acetic Anhydride/Lutidine for biosynthesis, VWR Chemicals BDH®

Supplier: VWR International
Danger

CB3754-112 CB3754-032 CB3754-001 CB3754-130 CB3754-157 CB3754-014
BDH0611-4LCS 4240.27 USD InStock
BDH0611-4L BDH0611-200L BDH0611-450ML BDH0611-20L BDH0611-2.5L BDH0611-56L
Cap B 50:20:30 Anhydrous ACN/Acetic Anhydride/Lutidine for biosynthesis, VWR Chemicals BDH®
Capping Reagent
BDH Biosynthesis Grade Reagents are designed for Biosynthesis research and production. Sizing available to fit your needs, from benchtop synthesizers to bulk production.

  • Custom Blends and Bulk Sizes available upon request.


Storage Temperature: Ambient
UN: 2924
ADR: 3 (8),II
Order Now
Specification Test Results
Description (Clear liquid free of suspended matter) Pass
Acetic Anhydride 19.0 - 21.0%
2,6-Lutidine 29.0 - 31.0%
Acetonitrile 49 - 51.0%
Color (at time of packaging) Max 10 APHA
Water Max 50 ppm
Acetic Anhydride (component purity) 99.5%
2,6-Lutidine (component purity) 99.2%
Acetonitrile (component purity) 99.9%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty