CB3970-001
CB3970-032
CB3970-112
CB3970-051
CB3970-014
CB3970-157
BDH0612-450MLCS
803.15
USD
InStock
BDH0612-450ML
BDH0612-4L
BDH0612-200L
BDH0612-56L
BDH0612-20L
BDH0612-2.5L
Cap A 8:1:1 THF/Lutidine/Acetic Anhydride for biosynthesis, VWR Chemicals BDH®
Capping Reagent
BDH Biosynthesis grade reagents are designed for biosynthesis research and production. Sizing available to fit your needs, from benchtop synthesizers to bulk production.
- Custom blends and bulk sizes available upon request.