Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cap A 8:1:1 THF/Pyridine/Acetic Anhydride for biosynthesis, VWR Chemicals BDH®

Supplier: VWR International
Danger

CB3975-051 CB3975-157 CB3975-014 CB3975-032 CB3975-001 CB3975-112
BDH0613-2.5LCS 1025.44 USD InStock
BDH0613-2.5L BDH0613-450ML BDH0613-4L BDH0613-200L BDH0613-56L BDH0613-20L
Cap A 8:1:1 THF/Pyridine/Acetic Anhydride for biosynthesis, VWR Chemicals BDH®
Capping Reagent
BDH Biosynthesis Grade Reagents are designed for Biosynthesis research and production. Sizing available to fit your needs, from benchtop synthesizers to bulk production.

  • Custom Blends and Bulk Sizes available upon request.


Storage Temperature: Ambient
UN: 2924
ADR: 3 (8),II
Order Now
Specification Test Results
Description (Clear liquid free of suspended matter) Pass
Acetic Anhydride 9.0 - 11.0%
Pyridine 9.0 - 11.0%
Tetrahydrofuran 79 - 81.0%
Color (at time of packaging) Max 10 APHA
Water Max 150 ppm
Acetic Anhydride (component purity) 99.5%
Pyridine (component purity) 99.8%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty