Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetonitrile-[D₃] (99.8% D) for NMR spectroscopy

Supplier: VWR International
Danger
Synonyms: Acetonitrile-d3Cyanomethane-D3Ethannitrile-D3Methyl-d3 cyanideAcetonitrile-[²H₃]Methyl cyanide-D3Trideuteroacetonitrile

300486 300836 301176 300542 300543
BDH0902-10MLEA 67.42 USD InStock
BDH0902-10ML BDH0902-25ML BDH0902-0.75ML BDH0902-100ML BDH0902-0.6ML
Acetonitrile-[D₃] (99.8% D) for NMR spectroscopy
Acetonitrile-[D3]
Deuterium Labeled Compounds. These high-quality products are used as solvents in NMR spectroscopy, R&D, tracers, and building blocks in pharmaceutical ingredients, as well as for the synthesis of organic light-emitting diodes (OLED).

  • Highest quality deuterated stable isotopes for NMR. NMR solvents are batch tested for both chemical and isotopic purity by 1H-NMR, GC and Karl Fischer titration.


Formula: D₃CCN
MW: 44.03 g/mol
Boiling Pt: 81.7 °C (1013 hPa)
Melting Pt: –45.7 °C
Density: 0.84 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001881
CAS Number: 2206-26-0
EINECS: 218-616-5
UN: 1648
ADR: 3,II
Order Now
Specification Test Results
Appearance/color Colorless
Appearance/form Liquid
D content 1H-NMR min. 99.80 %(mol)
w(H2O) 1H-NMR max. 0.020 %(w)
1H-NMR Purity conform
H2O + D2O Content by KF Coulometer max 0.030 %(w)
GC Content min. 99.0 %(w)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty