Synonyms:
Acetonitrile-d3
, Cyanomethane-D3
, Ethannitrile-D3
, Methyl-d3 cyanide
, Acetonitrile-[²H₃]
, Methyl cyanide-D3
, Trideuteroacetonitrile
300486
300836
301176
300542
300543
BDH0902-10MLEA
67.42
USD
InStock
BDH0902-10ML
BDH0902-25ML
BDH0902-0.75ML
BDH0902-100ML
BDH0902-0.6ML
Acetonitrile-[D₃] (99.8% D) for NMR spectroscopy
Acetonitrile-[D3]
Deuterium Labeled Compounds. These high-quality products are used as solvents in NMR spectroscopy, R&D, tracers, and building blocks in pharmaceutical ingredients, as well as for the synthesis of organic light-emitting diodes (OLED).
- Highest quality deuterated stable isotopes for NMR. NMR solvents are batch tested for both chemical and isotopic purity by 1H-NMR, GC and Karl Fischer titration.