Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Deuterium oxide (99.9% D) for NMR spectroscopy

Supplier: VWR International
Synonyms: Heavy waterWater-D2

301400 301689 300368 301496 301935 301401 301485
BDH0906-25MLEA 74.78 USD InStock
BDH0906-25ML BDH0906-1000ML BDH0906-0.75ML BDH0906-0.6ML BDH0906-10ML BDH0906-500ML BDH0906-100ML
Deuterium oxide (99.9% D) for NMR spectroscopy
Deuterium oxide
Deuterium Labeled Compounds. These high-quality products are used as solvents in NMR spectroscopy, R&D, tracers, and building blocks in pharmaceutical ingredients, as well as for the synthesis of organic light-emitting diodes (OLED).

  • Highest quality deuterated stable isotopes for NMR. NMR solvents are batch tested for both chemical and isotopic purity by 1H-NMR, GC and Karl Fischer titration.


Formula: D₂O
MW: 20 g/mol
Boiling Pt: 101 °C
Melting Pt: 3.81 °C
Density: 1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00044636
CAS Number: 7789-20-0
EINECS: 232-148-9
Order Now
Specification Test Results
Appearance/color Colorless
Appearance/form Liquid
D content 1H-NMR min. 99.90 %(mol)
1H-NMR Purity conform
Tritium max.74 Bq/g

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty