Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimethyl sulfoxide-[D₆] (99.95% D) for NMR spectroscopy

Supplier: VWR International
Synonyms: Dimethyl sulfoxide-[²H₆]Hexadeuterodimethyl sulfoxideDMSO-D6DMSO-d6Dimethyl sulphoxide-d6Dimethyl sulfoxide-d6Dimethyl sulphoxide-[²H₆]Hexadeuterodimethyl sulphoxide

301686 301296 302023 300374 300496
BDH0911-0.6MLEA 65.01 USD InStock
BDH0911-0.6ML BDH0911-10ML BDH0911-0.75ML BDH0911-100ML BDH0911-25ML
Dimethyl sulfoxide-[D₆] (99.95% D) for NMR spectroscopy
Dimethyl sulphoxide-[D6]
Deuterium Labeled Compounds. These high-quality products are used as solvents in NMR spectroscopy, R&D, tracers, and building blocks in pharmaceutical ingredients, as well as for the synthesis of organic light-emitting diodes (OLED).

  • Highest quality deuterated stable isotopes for NMR. NMR solvents are batch tested for both chemical and isotopic purity by 1H-NMR, GC and Karl Fischer titration.


Formula: D₃CS(O)CD₃
MW: 84.09 g/mol
Boiling Pt: ≥173 … ≤209 °C (1,013 mbar)
Melting Pt: ≥18 … ≤25 °C (1,013 mbar)
Density: 1.19 g/cm³ (20 °C)
Flash Pt: 88 °C (1 atm) (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002090
CAS Number: 2206-27-1
EINECS: 218-617-0
Order Now
Specification Test Results
Appearance/color Colorless
Appearance/form Liquid
D content 1H-NMR min. 99.95 %(mol)
w(H2O) 1H-NMR max. 0.010 %(w)
1H-NMR Purity conform
H2O + D2O Content by KF Coulometer max 0.020 %(w)
GC Content min. 99.5 %(w)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty