Synonyms:
Dimethyl sulfoxide-[²H₆]
, Hexadeuterodimethyl sulfoxide
, DMSO-D6
, DMSO-d6
, Dimethyl sulphoxide-d6
, Dimethyl sulfoxide-d6
, Dimethyl sulphoxide-[²H₆]
, Hexadeuterodimethyl sulphoxide
301686
301296
302023
300374
300496
BDH0911-0.6MLEA
65.01
USD
InStock
BDH0911-0.6ML
BDH0911-10ML
BDH0911-0.75ML
BDH0911-100ML
BDH0911-25ML
Dimethyl sulfoxide-[D₆] (99.95% D) for NMR spectroscopy
Dimethyl sulphoxide-[D6]
Deuterium Labeled Compounds. These high-quality products are used as solvents in NMR spectroscopy, R&D, tracers, and building blocks in pharmaceutical ingredients, as well as for the synthesis of organic light-emitting diodes (OLED).
- Highest quality deuterated stable isotopes for NMR. NMR solvents are batch tested for both chemical and isotopic purity by 1H-NMR, GC and Karl Fischer titration.