Synonyms:
Carbinol-D4
, Methyl alcohol-D4
, Methanol-[²H₄]
, Methyl-D3 alcohol-D
, Wood alcohol-D4
, Methanol-d4
, Hydroxy methane-D4
, Tetradeuteromethanol
300375
300504
300709
300710
302024
300503
BDH0913-0.75MLEA
45.21
USD
InStock
BDH0913-0.75ML
BDH0913-100ML
BDH0913-500ML
BDH0913-25ML
BDH0913-0.6ML
BDH0913-10ML
Methanol-[D₄] (99.8% D) for NMR spectroscopy
Methanol-[D4]
Deuterium Labeled Compounds. These high-quality products are used as solvents in NMR spectroscopy, R&D, tracers, and building blocks in pharmaceutical ingredients, as well as for the synthesis of organic light-emitting diodes (OLED).
- Highest quality deuterated stable isotopes for NMR. NMR solvents are batch tested for both chemical and isotopic purity by 1H-NMR, GC and Karl Fischer titration.