Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N-Dimethylformamide ≥99.8% ACS, VWR Chemicals BDH®

Supplier: VWR International
Danger
Synonyms: NSC 5356Formic acid dimethylamideDimethylformamideDMFADMFFormyldimethylamineN-Formyldimethylamine

BDHVBDH1117-4LG BDHVBDH1117-4LP BDHVBDH1117-204L
BDH1117-4LGCS 1105.19 USD InStock
BDH1117-4LG BDH1117-204L BDH1117-4LP
N,N-Dimethylformamide ≥99.8% ACS, VWR Chemicals BDH®
N,N-Dimethylformamide
Clear liquid.


Formula: HCON(CH₃)₂
MW: 73.09 g/mol
Boiling Pt: 153 °C (1013 hPa)
Melting Pt: –61 °C
Density: 0.949 g/cm³ (20 °C)
Flash Pt: 57.5 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
REACH: 01-2119475605-32
Merck Index: 13,03269
https://us-legacy.vwr.com/stibo/web/std.lang.all/25/06/2302506.jpg https://us-legacy.vwr.com/stibo/web/std.lang.all/06/12/16640612.jpg
Order Now
Specification Test Results
Appearance Clear, colorless liquid (can be slightly yellow)
Water (K.F.) Less than 0.15%
Residue Less than 0.005% (0.0010 g/21 mL)
GC Purity Greater than 99.8%
Color (APHA) Less than 10
Titrable acid Less than 0.5 meq/g (Not more than 1.0 mL of 0.01N sodium hydroxide)
Titrable base Less than 3 meq/g (Not more than 3.0 mL of 0.01N sodium hydroxide)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty