Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethanol denatured ACS (denatured with 5% isopropanol and 5% methanol), VWR Chemicals BDH®

Supplier: VWR International

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Ethyl alcohol denatured

BDHVBDH1158-19L BDHVBDH1158-4LP
BDH1158-4LPCS 840.89 USD InStock
BDH1158-4LP BDH1158-19L
Ethanol denatured ACS (denatured with 5% isopropanol and 5% methanol), VWR Chemicals BDH®
Ethanol denatured
  • Total alcohol: 95%
  • Ethyl alcohol: 85-87%
  • Denaturant: 4-5% methanol and 4.5–5.5% 2-propanol.

Meets ACS specifications for general use.

Ordering information: The purchase of Reagent Alcohol in containers larger than 4 liters requires a Letterhead Permit approval by the United States Treasury Tax and Trade Bureau (TTB).


Formula: H₃CCH₂OH
MW: 46.07 g/mol
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1170
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear, colorless liquid
Water 4.0–6.0%
Residue Less than 0.001% (0.0010g/122mL)
GC Purity Ethanol 85.0–87.0%
GC Purity Methanol 4.5–5.0%
GC Purity Isopropyl Alcohol 4.5–5.5%
Color (APHA) Less than 10
Specific gravity @ 20ºC 0.806-0.813
Acidity Less than 0.0025%

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty