Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®

Supplier: VWR International
Synonyms: GlysaninGlycerinGlyceritolPropane-1,2,3-triolOsmoglynPropanetriolGlycerolGlyrolGlycyl alcohol1,2,3-PropanetriolTrihydroxypropane

BDHVBDH1172-4LG BDHVBDH1172-19L BDHVBDH1172-1LP BDHVBDH1172-4LP
BDH1172-4LGCS 2156.38 USD InStock
BDH1172-4LG BDH1172-19L BDH1172-1LP BDH1172-4LP
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Glycerine
Clear liquid. Suitable for organic synthesis, cleaning, and prep scale synthesis.


Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Boiling Pt: 290 °C (1013 hPa)
Melting Pt: 18.6 °C
Density: 1.26 g/cm³ (20 °C)
Flash Pt: 177 °C
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
EINECS: 200-289-5
Merck Index: 13,04497
https://us-legacy.vwr.com/stibo/web/std.lang.all/47/74/734774.jpg
Order Now
Specification Test Results
Appearance Clear
Identification Passes test
Water (K.F.) Less than 0.3%
Residue after Ignition Less than 0.01%
Assay (calc from Specific Gravity, use % Glycerine test) Greater than 99.7%
Color (APHA) Less than 20
Specific Gravity (25ºC/25ºC) 1.2613 or greater
Chlorides Less than 10 ppm
Sulfates Less than 20 ppm
Heavy Metals (as Pb) Less than 5 ppm
Chlorinated Compounds (as Cl) Less than 30 ppm
Fatty Acids and Esters Passes test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty