Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Propanol ≥70%, Production Grade, VWR Chemicals BDH®

Supplier: VWR International
Danger
Synonyms: Isopropyl alcoholIsopropyl Rubbing Alcoholsec-PropanolIsopropyl rubbing alcohol2-Hydroxy propanesec-Propyl AlcoholDimethyl carbinol2-HydroxypropanePropane, 2-hydroxy-Propan-2-olIPAIsopropanol

BDHVBDH2014-1GLP BDHVBDH2014-55GLP BDHVBDH2014-5GLP 002V-08
BDH2014-5GLMSEA 464.78 USD InStock
BDH2014-5GLMS BDH2014-5GLP BDH2014-55GLP BDH2014-1GLP
2-Propanol ≥70%, Production Grade, VWR Chemicals BDH®
2-Propanol
Clear, colorless liquid.


Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Density: 0.8…0.92 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1987
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/19/69/731969.jpg
Order Now
Specification Test Results
Appearance Clear, Colorless
Specific Gravity at 60°F (0.872 – 0.883) g/mL

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty