Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Propanol ≥99.5% ACS, Production Grade, VWR Chemicals BDH®

Supplier: VWR International
Danger
Synonyms: Isopropyl alcoholIsopropyl Rubbing Alcoholsec-PropanolIsopropyl rubbing alcohol2-Hydroxy propanesec-Propyl AlcoholDimethyl carbinol2-HydroxypropanePropane, 2-hydroxy-Propan-2-olIPAIsopropanol

Filtered through a 0.2 µm filter.

BDHVBDH2016-1GLP BDHVBDH2016-55GLP BDHVBDH2016-5GLP BDHVBDH2016-55GLM
BDH2016-55GLPEA 2602.03 USD InStock
BDH2016-55GLP BDH2016-55GLM BDH2016-1GLP BDH2016-5GLP
2-Propanol ≥99.5% ACS, Production Grade, VWR Chemicals BDH®
2-Propanol
Soluble in water. Meets ACS specifications.


Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82 °C (1013 hPa)
Melting Pt: –89 °C
Density: 0.786 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://us-legacy.vwr.com/stibo/web/std.lang.all/19/69/731969.jpg
Order Now
Specification Test Results
GC Purity (CH3CHOHCH3) 99.5% Min.
Solubility in Water TO PASS TEST
Carbonyl Compounds: Acetone 0.002% Max.
Carbonyl Compounds: Propionaldehyde 0.002% Max.
Color (APHA) 10 APHA Max.
Residue upon Evaporation 0.001% Max.
Titrable Acid (µeq/g) 0.1 µeq/g Max.
Titrable Base (µeq/g) 0.1 µeq/g Max.
Water 0.2% Max.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty