Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Orthophosphoric acid ≥85% (w/w), ARISTAR®, Food Grade, VWR Chemicals BDH®

Supplier: BDH
Danger
Synonyms: Phosphoric acid

404200-22001 404200-03901 404200-03901
BDH3066-188LEA 11112.12 USD InStock
BDH3066-188L CABDH306219L BDH3062-19L
Orthophosphoric acid ≥85% (w/w), ARISTAR®, Food Grade, VWR Chemicals BDH®
Orthophosphoric acid


Formula: H₃PO₄
MW: 98 g/mol
Melting Pt: 28 °C
Density: 1.71…1.87 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear, colorless liquid
Assay (H3PO4) 85.0 - 88.0% (W/W)
Arsenic (As) 3 mg/kg max.
Cadmium (Cd) 3 mg/kg max.
Fluoride (F) 10 mg/kg max.
Identification (PO4) To Pass Test
Lead (Pb) 3 mg/kg max.
Manufacture Date Record Actual
Use by Date Record Actual

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty