Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetic acid glacial 99.7-100.5% ACS, meets analytical specification of USP, FCC, VWR Chemicals BDH®

Supplier: VWR International
Danger
Synonyms: Methanecarboxylic acidVinegar acidEthanoic acid

005700-80601 005700-03901 005700-03901 005700-80604 005700-80401 005700-80604 005700-80601 005700-80602 005700-80602 005700-80603 005700-80401 005700-80603 005700-22001
BDH3094-2.5LGCS 903.92 USD InStock
CABDH30942.5LG BDH3094-2.5LG CABDH3092500ML CABDH30983.8LP BDH3093-2.2LP CABDH30962.5L BDH3092-500MLP CABDH3100-19L CABDH3093-2.2P BDH3098-3.8LP BDH3096-2.5LPC BDH3100-19L BDH3102-201L
Acetic acid glacial 99.7-100.5% ACS, meets analytical specification of USP, FCC, VWR Chemicals BDH®
Acetic acid


Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118 °C (1013 hPa)
Melting Pt: 17 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 38.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8,II
REACH: 01-2119475328-30
https://us-legacy.vwr.com/stibo/web/std.lang.all/31/15/723115.jpg
Order Now
Specification Test Results
Assay (CH3COOH) 99.7% min.(ACS)/99.5-100.5%(USP/FCC)
Acetic Anhydride 0.01% max.(ACS)
Chloride (as Cl) To Pass Test(USP)
Chloride (Cl) 1 ppm max.(ACS)
Color (APHA) 10 max.(ACS)
Congealing Temperature 15.6 C min.(USP/FCC)
Dilution Test To pass test(ACS)
Heavy Metals (as Pb) 0.5 ppm max.(ACS)
Identification: acetate To Pass Test(USP/FCC)
Iron (Fe) 0.2 ppm max.(ACS)
Lead (Pb) 0.5 mg/kg max.(FCC)
Nonvolatile residue 1.0mg max.(USP)/0.005% max.(FCC)
Oxidizable substances To pass test(USP/FCC)
Residue After Evaporation 0.001% max.(ACS)
Substances Reducing Dichromate To pass test (ACS)
Substances Reducing Permanganate To pass test (ACS)
Sulfate 1 ppm max.(ACS)
Sulfate (SO4) To Pass Test(USP)
Titrable Base 0.0004 meq/g max.(ACS)
Manufacture Date Record Actual
Expiration date Record Actual

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty