Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Orthophosphoric acid 85% ACS, meets analytical specification of NF, VWR Chemicals BDH®

Supplier: BDH
Danger
Synonyms: Phosphoric acid

408000-80401 408000-80401
BDH3118-3.8LPCS 969.32 USD InStock
BDH3118-3.8LP CABDH3118-3.8
Orthophosphoric acid 85% ACS, meets analytical specification of NF, VWR Chemicals BDH®
Orthophosphoric acid


Formula: H₃PO₄
MW: 98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8,III
Order Now
Specification Test Results
Assay (H3PO4) 85.0-88.0%
Alkali phosphates To Pass test
Identification: NF To Pass test
Limit of Nitrate To Pass test
Phosphorous or hypophosphorous acid To Pass test
Sulfate To Pass test
Manufacture Date Record Actual
Expiration Date Record Actual

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty