Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 36.5 - 38.0% ACS, VWR Chemicals BDH®

Supplier: VWR International

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Chlorohydric acidHydrochloric acid gasHydrochlorideHydrogen chloride gasMuriatic acid

257900-80401 257900-80601 257900-21601 257900-80606 257900-80603 257900-80601 257900-80606 257900-22001 257900-80602 257900-80401 257900-80603 257900-21601 257900-22001 257900-80602 257900-21801
BDH3038-191LEA 3992.64 USD InStock
CABDH3128-2.2P BDH3038-191L BDH3028-2.5LG BDH3026-500MLP BDH3128-2.2LP CABDH3030PC BDH3034-19L CABDH30323.8LP CABDH303419L BDH3032-3.8LP CABDH30282.5LG BDH3036-54L CABDH3026P CABDH3038191L BDH3030-2.5LPC
Hydrochloric acid 36.5 - 38.0% ACS, VWR Chemicals BDH®
Hydrochloric acid


Formula: HCl
MW: 36.46 g/mol
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: –30 °C
Density: 1.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Assay (HCl) 36.5-38.0%
Assay (HCl) Be' 22.7 deg Be' min.
Ammonium (NH4) 0.0003% max.
Appearance Clear, colorless fuming liquid
Arsenic (As) 10 ppb max.
Bromide or Iodide To pass test
Clarity of Solution Free from suspended matter/sediment
Color (APHA) 10 max.
Free Bromine or Chlorine To pass test
Free Chlorine (Cl2) 0.0001% max.
Heavy Metals (as Pb) 0.0001% max.
Identification FCC To pass test
Identification NF To pass test
Iron (Fe) 0.00002% max
Lead (Pb) FCC/USP 1 mg/kg max.
Mercury FCC 0.10 ppm max.
Organic Compounds - Benzene 0.05 ppm max.
Organic Compounds - Total fluorinated organic compounds 25 ppm max.
Reducing Substances To pass test
Residue After Ignition 0.0005% max.
Specific Gravity at 60°F Record only (approx. 1.17-1.20)
Sulfate (SO4) 0.0001% max.
Sulfite (SO3) 0.0001% max.
Total Organic Compounds 5 ppm max.

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty