Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Urea 99.0-100.5%, crystallised ACS, VWR Chemicals BDH®

Supplier: VWR International
Synonyms: Carbonyl diamideCarbamideCarbonyldiamideDiaminomethanone

BDHVBDH4602-500G
BDH4602-500GCS 635.77 USD InStock
BDH4602-500G
Urea 99.0-100.5%, crystallised ACS, VWR Chemicals BDH®
Urea


Formula: NH₂CONH₂
MW: 60.06 g/mol
Boiling Pt: 197 °C (1013 hPa)
Melting Pt: 132.7 °C
Density: 1.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008022
CAS Number: 57-13-6
EINECS: 200-315-5
REACH: 01-2119463277-33
Merck Index: 12,10005
https://us-legacy.vwr.com/stibo/web/std.lang.all/70/46/2297046.jpg
Order Now
Specification Test Results
Assay 99.0 - 100.5 %
Melting point (132-135°C) Passes test
Heavy metals (as Pb) Max. 0.001 %
Insoluble matter Max. 0.01 %
Residue after ignition Max. 0.01 %
Cl (Chloride) Max. 5 ppm
SO4 (Sulphate) Max. 0.001 %
Fe (Iron) Max. 0.001 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty