Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Starch 1% (w/v) in aqueous solution indicator, VWR Chemicals BDH®

Supplier: VWR International

BDH51040-1A BDH51040-4A
BDH5104-1LEA 52.01 USD InStock
BDH5104-1L BDH5104-4L
Starch 1% (w/v) in aqueous solution indicator, VWR Chemicals BDH®
Starch
This indicator is prepared using soluble starch and high purity water. Salicylic acid is added as a preservative. Store at 5 to 30 °C tightly closed in the original containers.


Formula: (C₆H₁₀O₅)n
MW: 162.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-25-8
EINECS: 232-679-6
Order Now
Specification Test Results
Appearance Translucent white liquid
Suitability for use Colorless (Iodine absent) - Blue (Iodine present)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty