Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Starch 2% (w/v) in aqueous solution indicator, VWR Chemicals BDH®

Supplier: VWR International

BDH51060-500A
BDH5106-500MLEA 49.74 USD InStock
BDH5106-500ML
Starch 2% (w/v) in aqueous solution indicator, VWR Chemicals BDH®
Starch
Starch indicator, 2% (w/v) aqueous solution, mercury free, for iodometric titrations.


Formula: (C₆H₁₀O₅)n
MW: 162.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-25-8
EINECS: 232-679-6
Order Now
Specification Test Results
Appearance Translucent white liquid
Suitability for use Colorless (Iodine absent) - Blue (Iodine present)
Storage condition 5-30 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty