Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrogen peroxide 6% in aqueous solution stabilized, VWR Chemicals BDH®

Supplier: VWR International
Danger

BDHVBDH7103-2
BDH7103-2CS 921.6 USD InStock
BDH7103-2
Hydrogen peroxide 6% in aqueous solution stabilized, VWR Chemicals BDH®
Hydrogen peroxide
Made with purified water.


Formula: H₂O₂
MW: 34.01 g/mol
Density: 1.022 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011333
CAS Number: 7722-84-1
EINECS: 231-765-0
Order Now
Specification Test Results
Assay 5.80 - 6.40 %
Appearance/color Clear liquid
Stabilizer (acetanilide) Passes test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty