Synonyms:
Chlorohydric acidHydrochloric acid gasHydrochlorideHydrogen chloride gasMuriatic acid
BDHVBDH7200-1
BDHVBDH7200-4
BDHVBDH7200-7
BDH7200-1CS
489.95
USD
InStock
BDH7200-1
BDH7200-7
BDH7200-4
Hydrochloric acid 0.10 N, VWR Chemicals BDH®
Hydrochloric acid
Meets AOAC 936.15 standard and ASTM E200-91 standard. Made with high purity acid and deionized water. Suitable as a titrant.