Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iodine solution 0.1 N, VWR Chemicals BDH®

Supplier: VWR International

BDHVBDH7207-1 BDHVBDH7207-4
BDH7207-4CS 939.09 USD InStock
BDH7207-4 BDH7207-1
Iodine solution 0.1 N, VWR Chemicals BDH®
Iodine
Made with iodine crystals, potassium iodide and deionized water.


Formula: I₂
MW: 253.81 g/mol
MDL Number: MFCD00011355
CAS Number: 7553-56-2
UN: 3495
ADR: 8,III
Order Now
Specification Test Results
Normality 0.0995 - 0.1005 N
Appearance/color Dark brown liquid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty