Synonyms:
Caustic sodaLyeSodium hydrateSoda lye
BDHVBDH7221-7
BDHVBDH7221-4
BDHVBDH7221-1
BDH7221-4CS
380.35
USD
InStock
BDH7221-4
BDH7221-1
BDH7221-7
Sodium hydroxide 0.5 N in aqueous solution, VWR Chemicals BDH®
Sodium hydroxide
AOAC, APHA, and ASTM. Suitable as titrant.