Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 1.0 N in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International
Danger
Synonyms: Caustic sodaLyeSodium hydrateSoda lye

BDHVBDH7222-4 BDHVBDH7222-7 BDHVBDH7222-1
BDH7222-1CS 546.13 USD InStock
BDH7222-1 BDH7222-7 BDH7222-4
Sodium hydroxide 1.0 N in aqueous solution, VWR Chemicals BDH®
Sodium hydroxide
NIST traceable. Suitable as a titrant.


Formula: NaOH
MW: 40 g/mol
Density: 1.04 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 3266
ADR: 8,III
Order Now
Specification Test Results
Appearance/color Clear liquid
Traceable to NIST SRM 84 Yes
Normality 0.995 - 1.005 N
Shelf-life 2 years
Manufacture date As reported
Expiration date As reported

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty