Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium thiosulfate 0.025 N in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International
Synonyms: di-Sodium thiosulfateSodium hyposulphitedi-Sodium thiosulphateSodiumthiosulfateSodium hyposulfiteAntichlorSodiumthiosulphate

BDHVBDH7227-1 BDHVBDH7227-4
BDH7227-4CS 453.42 USD InStock
BDH7227-4 BDH7227-1
Sodium thiosulfate 0.025 N in aqueous solution, VWR Chemicals BDH®
Sodium thiosulphate
Suitable as a titrant. Made with deionized water.


Formula: Na₂S₂O₃
MW: 158.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
EINECS: 231-867-5
Order Now
Specification Test Results
Appearance/color Clear liquid
N.I.S.T. traceable to SRM 136 Yes
Normality 0.0245 - 0.0255 N
Shelf-life 6 months

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty