Synonyms:
Hydrogen sulfateOil of vitriolVitriol brown oil
BDHVBDH7232-1
BDHVBDH7232-4
BDH7232-1CS
474.15
USD
InStock
BDH7232-1
BDH7232-4
Sulfuric acid 1.0 N in aqueous solution, VWR Chemicals BDH®
Sulfuric acid
APHA. Made with high purity acid and deionized water. Suitable as a titrant.