Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Barium chloride 10% (w/v) in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International
Warning

BDHVBDH7238-1
BDH7238-1CS 867.93 USD InStock
BDH7238-1
Barium chloride 10% (w/v) in aqueous solution, VWR Chemicals BDH®
Barium chloride
Made with deionized water.


Formula: BaCl₂
MW: 208.24 g/mol
Boiling Pt: ∼100 °C (1013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00003445
CAS Number: 10361-37-2
EINECS: 233-788-1
UN: 3287
ADR: 6.1,III
Order Now
Specification Test Results
Appearance/color Clear to slightly turbid liquid
Shelf-life 1 year
N.I.S.T. traceable to SRM 919 Yes
Normality 9.50 - 10.50 N

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty