Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 10.0 N in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

BDHVBDH7247-1 BDHVBDH7247-7 BDHVBDH7247-5 BDHVBDH7247-10 BDHVBDH7247-4
BDH7247-1CS 597.94 USD InStock
BDH7247-1 BDH7247-4 BDH7247-5 BDH7247-7 BDH7247-10
Sodium hydroxide 10.0 N in aqueous solution, VWR Chemicals BDH®
Sodium hydroxide
Made with deionized water. Suitable as a titrant.

Shelf-life 2 years.


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 109 °C (1013 hPa)
Density: 1.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II
Order Now
Specification Test Results
Normality 9.9 - 10.1 N
Appearance Clear liquid
Traceable to NIST SRM 84 Confirmed
Prepared to formulation on file Confirmed

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty