Would you like to log in again?
Acetate Buffer Solution, pH 4.0, VWR Chemicals BDH®
Supplier: Avantor
BDH®
BDHVBDH7300-7 BDHVBDH7300-1 BDHVBDH7300-4
BDH7300-1CS 735.18 USD InStock
BDH7300-1 BDH7300-7 BDH7300-4
Acetate Buffer Solution, pH 4.0, VWR Chemicals BDH®
Buffers pH Calibration Buffers
APHA 4500-C1 and ASTM 1253-91 for chlorine.
Standardized at 25 °C against chemicals whose certification is traceable to NIST.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty