Synonyms:
Di-Sodium Ethylenediaminetetraacetic acidDisodium EDTA
BDHVBDH7314-10
BDHVBDH7314-1
BDHVBDH7314-4
BDH7314-4CS
643.13
USD
InStock
BDH7314-4
BDH7314-1
BDH7314-10
EDTA disodium salt 0.100 M (0.200 N) in aqueous solution, VWR Chemicals BDH®
EDTA disodium salt
Suitable as a titrant. Made with deionized water.