Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium iron (II) sulfate 0.1 N in dil. sulfuric acid, VWR Chemicals BDH®

Supplier: VWR International
Synonyms: di-Ammonium iron (II) sulfateAmmonium ferrous sulphateAmmonium ferrous sulfatedi-Ammonium iron (II) sulphate

BDHVBDH7320-1
BDH7320-1CS 1609.26 USD InStock
BDH7320-1
Ammonium iron (II) sulfate 0.1 N in dil. sulfuric acid, VWR Chemicals BDH®
Ammonium iron (II) sulphate
Made with deionized water and high purity sulfuric acid.


Formula: H₈FeN₂O₈S₂
MW: 284.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00051039
CAS Number: 10045-89-3
EINECS: 233-151-8
Order Now
Specification Test Results
Appearance/color Pale blue-green liquid
Normality 0.0995 - 0.1005 N
Shelf-life 6 months

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty