Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mineral oil, white (petroleum), VWR Chemicals BDH®

Supplier: VWR International
Warning
Synonyms: Matrix oilMineral oilParaffin oil

BDHVBDH7338-4
BDH7338-4CS 645.67 USD InStock
BDH7338-4
Mineral oil, white (petroleum), VWR Chemicals BDH®
White mineral oil (petroleum)


Boiling Pt: 300 °C (1013 hPa)
Density: 0.872 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00131611
CAS Number: 8042-47-5
EINECS: 232-455-8
Order Now
Specification Test Results
Appearance Clear liquid
Odour description Confirmed
Specific gravity (25°C/25°C) 0.852 -- 0.876
Viscosity (100°F) (ASTM-D2161) (SUS) 180.0 -- 190.0
Viscosity (40°C) (ASTM-D445) 34.9 -- 37.3 cSt

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty