Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenolphthalein solution 0.5%, VWR Chemicals BDH®

Supplier: VWR International
Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one

BDHVBDH7342-2 BDHVBDH7342-1
BDH7342-1CS 618.42 USD InStock
BDH7342-1 BDH7342-2
Phenolphthalein solution 0.5%, VWR Chemicals BDH®
Phenolphthalein
Made with 50% isopropanol.


Formula: C₂₀H₁₄O₄
MW: 318.33 g/mol
MDL Number: MFCD00005913
CAS Number: 77-09-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/72/01/2297201.jpg
Order Now
Specification Test Results
Appearance/color Clear liquid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty