Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenolphthalein 1% in alcohol, VWR Chemicals BDH®

Supplier: VWR International
Danger
Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one

BDHVBDH7344-2 BDHVBDH7344-1 BDHVBDH7344-4
BDH7344-1CS 1487.08 USD InStock
BDH7344-1 BDH7344-2 BDH7344-4
Phenolphthalein 1% in alcohol, VWR Chemicals BDH®
Phenolphthalein
Made with denatured ethanol.


Formula: C₂₀H₁₄O₄
MW: 318.33 g/mol
Boiling Pt: ∼78 °C (1013 hPa)
Density: ∼0.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005913
CAS Number: 77-09-8
EINECS: 201-004-7
UN: 1170
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/72/01/2297201.jpg
Order Now
Specification Test Results
Appearance/color Clear liquid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty